Order Entry
Poland
ContactUsLinkComponent
recombinant protein, aof2 (human) recombinant protein (q01), human aof2 partial orf (np_055828, 753 a.a. - 851 a.a.) recombinant pro tein with gst-tag at n-terminal. 1 * 10 µG
Catalog # ABNOH00023028-Q01
Supplier:  Abnova
recombinant protein, aof2 (human) recombinant protein (q01), human aof2 partial orf (np_055828, 753 a.a. - 851 a.a.) recombinant pro tein with gst-tag at n-terminal. 1 * 10 µG
Catalog # ABNOH00023028-Q01
Supplier:  Abnova