Order Entry
Poland
Orders LinkContactUsLinkComponent
CHES, 0.5 M buffer solutions

CHES, 0.5 M buffer solutions

CHES buffer solution, 0,5 M, 9.0, 100 mL
Catalog # J63670.AE
CHES, 0.5 M buffer solutions
Catalog # J63670.AE
CAS Number:  
CHES, 0.5 M buffer solutions
CHES, 0.5 M buffer solutions

CHES buffer solution, 0,5 M, 9.0, 100 mL

CAS Number:  
Price
565 zł
Log in for availability
1 * 100 mL
Product Details & Documents

2-(Cyclohexylamino)ethanesulfonic acid, CHES.

CAS No.: 103-47-9
MDL No.: MFCD00003835

Specifications

Specifications

  • Concentration:
    0,5 M
  • Pk:
    100 mL
  • Description:
    CHES buffer solution
  • pH value:
    9.0
  • Cat. No.:
    J63670.AE
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00003835
  • CAS No.:
    103-47-9

Specifications