Succinate, 0,2 M buffer solution, pH 6,5
Succinate buffer solution, 0,2 M, 6,5, 500 mL
Supplier: Thermo Fisher Scientific
Succinate, 0,2 M buffer solution, pH 6,5
Catalog # J62078.AP
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
CAS No.: 150-90-3
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless
Odour: Odourless
MDL No.: MFCD00002789
Documents
Specifications
Specifications
- Concentration:0,2 M
- Pk:500 mL
- Description:Succinate buffer solution
- pH value:6,5
- Cat. No.:J62078.AP
- Odour/scent:Odourless
- Storage temperature:2°C to 8°C
- Physical form:Liquid
- MDL No.:MFCD00002789
- Appearance:Colourless
- CAS No.:150-90-3



