Order Entry
Poland
Orders LinkContactUsLinkComponent
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)

Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)

TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer), Glass bottle, 1 L
Catalog # J62449.K2
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
Catalog # J62449.K2
CAS Number:  
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)

TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer), Glass bottle, 1 L

CAS Number:  
Price
398 zł
Log in for availability
1 * 1 L
Product Details & Documents

Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00236358

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer)
  • Cat. No.:
    J62449.K2
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00236358
  • Appearance:
    Colourless

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.