Order Entry
Poland
Orders LinkContactUsLinkComponent
MOPS-EDTA-sodium acetate buffer (MESA)

MOPS-EDTA-sodium acetate buffer (MESA)

MOPS-EDTA-sodium acetate buffer (MESA), 500 mL
Catalog # J60184.AP
MOPS-EDTA-sodium acetate buffer (MESA)
Catalog # J60184.AP
CAS Number:  
MOPS-EDTA-sodium acetate buffer (MESA)
MOPS-EDTA-sodium acetate buffer (MESA)

MOPS-EDTA-sodium acetate buffer (MESA), 500 mL

CAS Number:  
Price
699 zł
Log in for availability
1 * 500 mL
Product Details & Documents

Physical Form: Liquid
MDL No.: MFCD00131625

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    MOPS-EDTA-sodium acetate buffer (MESA)
  • Cat. No.:
    J60184.AP
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00131625

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.