Order Entry
Poland
Orders LinkContactUsLinkComponent
HEPES, 0.5 M buffer solutions

HEPES, 0.5 M buffer solutions

HEPES buffer solution, 0,5 M, 8,0, Glass bottle, 100 mL
Catalog # J63002.AE
HEPES, 0.5 M buffer solutions
Catalog # J63002.AE
CAS Number:  
HEPES, 0.5 M buffer solutions
HEPES, 0.5 M buffer solutions

HEPES buffer solution, 0,5 M, 8,0, Glass bottle, 100 mL

CAS Number:  
Price
546 zł
Log in for availability
1 * 100 mL
Product Details & Documents
Specifications

Specifications

  • Concentration:
    0,5 M
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    HEPES buffer solution
  • pH value:
    8,0
  • Cat. No.:
    J63002.AE
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00006158
  • CAS No.:
    7365-45-9

Specifications