Order Entry
Poland
ContactUsLinkComponent
Free from toxic heavy metals in formulation. Manufactured to meet ASTM protocols. Tested, certified & batch traceable Density Standa rd 0.8098g/ml @ 80C 1 * 100 mL
Catalog # REGEDEN80110
Supplier:  REAGECON
Free from toxic heavy metals in formulation. Manufactured to meet ASTM protocols. Tested, certified & batch traceable Density Standa rd 0.8098g/ml @ 80C 1 * 100 mL
Catalog # REGEDEN80110
Supplier:  REAGECON
Frequently Bought Together