Order Entry
Poland
ContactUsLinkComponent
Sucrose Standard 40% - 1.39986 (± 0.00004) manufactured in accordance with ICUMSA guidelines, tested and certified to 2 decimals pla ce. Extended shelf-life of 12 weeks 1 * 15 mL
  REGEBS40
 :  REAGECON
Sucrose Standard 40% - 1.39986 (± 0.00004) manufactured in accordance with ICUMSA guidelines, tested and certified to 2 decimals pla ce. Extended shelf-life of 12 weeks 1 * 15 mL
  REGEBS40
 :  REAGECON
Frequently Bought Together