FERROIN INIE DOTYCZYICATOR 1/40MOLAR 1 * 50 mL
Catalog # CHMP224238538.0050
Supplier: CHEMPUR
Frequently Bought Together
Cyclopentyl methyl ether (CPME), EMPLURA®
Supplier: Merck
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
N,N-Dimethylformamide 99+%, extra pure
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.8% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylacetamide ≥99%
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
(±)-2-Butanol ≥99%
Supplier: Thermo Fisher Scientific
Diethyl ether ≥99.5% (by GC) stabilised, Puriss., meets analytical specification of BP, Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Ammonium chloride, pure analytical reagent
Supplier: POCH AVANTOR BRAND
Pyridine 99+% ACS
Supplier: Thermo Fisher Scientific
Petroleum spirit, 35…60 °C ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals