Burette 50:0.1ml CL.as with ns ptfeplug clear glass acc. to mohr,made of borosilicate glass 3.3 in compliance with din en iso 385 ve ry good chemical resistance minimal thermal expansion high resistance to thermal shocks 1 * 2 items
Catalog # MARI2422131
Dostawca: MARIENFELD
Frequently Bought Together