RECOMBINANT/NATIVE PROTEIN, MALTOSE BINDING PROTEIN (ESCHERICHIA COLI) RECOMBINANT PROTEIN, ESCHERICHIA COLI MALTOSE BINDING PROTEIN (1 A.A. - 387 A.A.) FULL-LENGTH RECOMBINANT PROTEIN EXPRESSED IN ESCHERICHIA COLI. 1 * 500 µG
Catalog # ABNOP4989
Dostawca: Abnova
Frequently Bought Together