Order Entry
Poland
ContactUsLinkComponent
KIT, LSD1 DEMETHYLASE ACTIVITY/INHIBITION ASSAY KIT (FLUOROMETRIC), LSD1 DEMETHYLASE ACTIVITY/INHIBITION ASSAY KIT (FLUOROMETRIC) IS USED FOR AN EASY AND FAST FLUOROMETRIC MEASUREMENT OF LSD1 ACTIVITY OR INHIBITION. 1 * 1 KIT
Catalog # ABNOKA1603
Supplier:  Abnova
KIT, LSD1 DEMETHYLASE ACTIVITY/INHIBITION ASSAY KIT (FLUOROMETRIC), LSD1 DEMETHYLASE ACTIVITY/INHIBITION ASSAY KIT (FLUOROMETRIC) IS USED FOR AN EASY AND FAST FLUOROMETRIC MEASUREMENT OF LSD1 ACTIVITY OR INHIBITION. 1 * 1 KIT
Catalog # ABNOKA1603
Supplier:  Abnova