Order Entry
Poland
Orders LinkContactUsLinkComponent
Guaiacol
Glass bottle for solids, 50 g
  CAYM70430-50
Guaiacol
  CAYM70430-50
 :  90-05-1
Guaiacol
Glass bottle for solids, 50 g
 :  90-05-1
148 zł
1 * 50 g
 :  1-Hydroxy-2-methoxybenzene, 2-Hydroxyanisole, 2-Methoxyphenol
  •  :90-05-1
  •  :C₇H₈O₂
  •  :MFCD00002185
  •  :124.14 g/mol
  •  :13,04568
  •  :205 °C (1013 hPa)
  •  :28…32 °C
  •  :1.1534 g/cm³ (0 °C)
  •  :82 °C
  •  :Ambient
  •  :201-964-7
  •  :2810
  •  :6.1,III
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g