Pterostilbene
Glass bottle for solids, 50 mg
Catalog # CAYM13000-50
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: (E)-4-(3,5-Dimethoxystyryl)phenol, 4-[(E)-2-(3,5-Dimethoxyphenyl)ethenyl]phenol
- CAS Number:537-42-8
- Formula:C₁₆H₁₆O₃
- MDL Number:MFCD00238710
- Molecular Weight:256.2964 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 mg



