E-64
Glass bottle for solids, 25 mg
Catalog # CAYM10007963-25
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 mg
- Cat. No.:CAYM10007963-25
- Molecular formula:C₁₅H₂₇N₅O₅
- Protein/peptide name:E-64
- Storage conditions:Freezer
- Protein synonyms:3S)-(+)-trans-Epoxy succinyl-L-leucylamido-4-guanidino butane)|((2S
- MDL No.:MFCD00080261
- CAS No.:66701-25-5



