E-64
Glass bottle for solids, 5 mg
Catalog # CAYM10007963-5
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- Cat. No.:CAYM10007963-5
- Molecular formula:C₁₅H₂₇N₅O₅
- Protein/peptide name:E-64
- Storage conditions:Freezer
- Protein synonyms:3S)-(+)-trans-Epoxy succinyl-L-leucylamido-4-guanidino butane)|((2S
- MDL No.:MFCD00080261
- CAS No.:66701-25-5
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



