Order Entry
Poland
Orders LinkContactUsLinkComponent
Cream butyrometer, without stopper
Cream butyrometer, without stopper
Butyrometer for cream, Roeder´s, 0 - 5 - 70 % : 1,0
Catalog # 614-0118
Supplier:   GERBER FUNKE
Cream butyrometer, without stopper
Catalog # 614-0118
Supplier:  GERBER FUNKE
Cream butyrometer, without stopper
Cream butyrometer, without stopper
Butyrometer for cream, Roeder´s, 0 - 5 - 70 % : 1,0
Price
719 zł
Log in for availability
1 * 10 SZT
Product Details & Documents
According to Roeder´s weighing method.
Specifications
  • Description:
    Butyrometer for cream, Roeder´s, 0 - 5 - 70 % : 1,0
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Weighing boat for butter for butyrometers acc. to Roeder's method
    Weighing boat for butter for butyrometers acc. to Roeder's method
    614-2034
    Log in for availability
    1 * 1 SZT - 22,10 zł
Product Information
  • Catalog #
    Availability
    Price
  • Cream beaker, unperforated, for ice cream and condensed milk butyrometer and cream butyrometer acc. to Roeder´s method
    Cream beaker, unperforated, for ice cream and condensed milk butyrometer and cream butyrometer acc. to Roeder´s method
    614-2032
    Log in for availability
    1 * 1 SZT - 8,30 zł
Product Information
  • Catalog #
    Availability
    Price
  • Stopper, rubber, with hole, for all weighing method butyrometers
    Stopper, rubber, with hole, for all weighing method butyrometers
    614-2030
    Log in for availability
    1 * 100 SZT - 306 zł
Product Information
  • Catalog #
    Availability
    Price
  • Stopper, rubber, for sealing the bulbs of all weighing method butyrometers
    Stopper, rubber, for sealing the bulbs of all weighing method butyrometers
    614-2029
    Log in for availability
    1 * 100 SZT - 102 zł