Methanol
Glass bottle, 1 L
CHMP616219908.1000
: CHEMPUR
: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
- :67-56-1
- :CH₃OH
- :MFCD00004595
- :32.04 g/mol
- :13,05984
- :64.6 °C (1013 hPa)
- :–98 °C
- :0.7918 g/cm³ (20 °C)
- :11 °C (closed cup)
- :Ambient
- :200-659-6
- :1230
- :3,II
- :01-2119433307-44
- Pack type:Glass bottle
- Pk:1 L
- Cat. No.:CHMP616219908.1000
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Environmentally Preferable:Safer Human and Environmental Health



