Order Entry
Poland
Orders LinkContactUsLinkComponent
Copper(II) carbonate basic, pure

Copper(II) carbonate basic, pure

500 g
Catalog # CHMP426586309.0500
Irritant
Supplier:   CHEMPUR
Copper(II) carbonate basic, pure
Catalog # CHMP426586309.0500
Supplier:  CHEMPUR
CAS Number:  12069-69-1
Copper(II) carbonate basic, pure

500 g

CAS Number:  12069-69-1
Price
153 zł
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Copper (II) carbonate dihydroxide, Copper(II) carbonate, basic, Basic copper carbonate, Copper (II) carbonate-copper (II) hydroxide (1:1)
  • CAS Number:12069-69-1
  • Formula:CuCO₃·Cu(OH)₂
  • MDL Number:MFCD00010976
  • Molecular Weight:221.12 g/mol
  • Merck Index:13,02658
  • Melting Pt:200 °C
  • Density:4 g/cm³ (20 °C)
  • EINECS:235-113-6
  • UN:3288
  • ADR:6.1,III
Specifications

Specifications

  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.