Copper(II) carbonate basic, pure
250 g
Catalog # CHMP426586309.0250
Supplier: CHEMPUR
Copper(II) carbonate basic, pure
Catalog # CHMP426586309.0250
Supplier: CHEMPUR
CAS Number: 12069-69-1
Jump to:
Product Details & Documents
Synonyms: Copper (II) carbonate dihydroxide, Copper(II) carbonate, basic, Basic copper carbonate, Copper (II) carbonate-copper (II) hydroxide (1:1)
- CAS Number:12069-69-1
- Formula:CuCO₃·Cu(OH)₂
- MDL Number:MFCD00010976
- Molecular Weight:221.12 g/mol
- Merck Index:13,02658
- Melting Pt:200 °C
- Density:4 g/cm³ (20 °C)
- EINECS:235-113-6
- UN:3288
- ADR:6.1,III
Specifications
Specifications
- Pk:250 g
- Cat. No.:CHMP426586309.0250



