Order Entry
Poland
Orders LinkContactUsLinkComponent
Zinc, pellets

Zinc, pellets

Glass bottle for solids, 100 g
Catalog # CHMP422631359.0100
Supplier:   CHEMPUR
Zinc, pellets
Catalog # CHMP422631359.0100
Supplier:  CHEMPUR
CAS Number:  7440-66-6
Zinc, pellets

Glass bottle for solids, 100 g

CAS Number:  7440-66-6
Price
331 zł
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:7440-66-6
  • Formula:Zn
  • MDL Number:MFCD00011291
  • Molecular Weight:65.39 g/mol
  • Merck Index:12,10255
  • Boiling Pt:908 °C (1013 hPa)
  • Melting Pt:420 °C
  • Density:7.14 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-175-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.