Order Entry
Poland
Orders LinkContactUsLinkComponent
Lead

Lead

Plastic bottle for solids, 500 g
Catalog # CHMP527034554.0500
Health
Irritant
Environment
Supplier:   CHEMPUR
Lead
Catalog # CHMP527034554.0500
Supplier:  CHEMPUR
CAS Number:  7439-92-1
Lead

Plastic bottle for solids, 500 g

CAS Number:  7439-92-1
Price
463 zł
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:7439-92-1
  • Formula:Pb
  • MDL Number:MFCD00134050
  • Molecular Weight:207.2 g/mol
  • Merck Index:12,05410
  • Boiling Pt:1740 °C (1013 hPa)
  • Melting Pt:327 °C
  • Density:11.3437 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-100-4
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.