Order Entry
Poland
Orders LinkContactUsLinkComponent
Lead | Avantor

Lead

Glass bottle for solids, 250 g
numer katalogowy CHMP527034554.0250
Health
Irritant
Environment
Dostawca:   CHEMPUR
Lead
numer katalogowy CHMP527034554.0250
Dostawca:  CHEMPUR
Numer CAS:  7439-92-1
Lead

Glass bottle for solids, 250 g

Numer CAS:  7439-92-1
Price
295 zł
Log in for availability
1 * 250 g
Product Details & Documents
  • CAS Number:7439-92-1
  • Formula:Pb
  • MDL Number:MFCD00134050
  • Molecular Weight:207.2 g/mol
  • Merck Index:12,05410
  • Boiling Pt:1740 °C (1013 hPa)
  • Melting Pt:327 °C
  • Density:11.3437 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-100-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications

Rekomendacje

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.