Copper
Glass bottle for solids, 100 g
CHMP526111851.0100
: CHEMPUR
- :7440-50-8
- :Cu
- :MFCD00010965
- :63.55 g/mol
- :13,02545
- :2595 °C (1013 hPa)
- :1083 °C
- :8.94 g/cm³ (20 °C)
- :Ambient
- :231-159-6
- Pack type:Glass bottle for solids
- Pk:100 g