Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Copper

Glass bottle for solids, 50 g
Catalog # CHMP526111851.0050
Supplier:   CHEMPUR
Copper
Catalog # CHMP526111851.0050
Supplier:  CHEMPUR
CAS Number:  7440-50-8
Copper

Glass bottle for solids, 50 g

CAS Number:  7440-50-8
Price
274 zł
Log in for availability
1 * 50 g
Product Details & Documents
  • CAS Number:7440-50-8
  • Formula:Cu
  • MDL Number:MFCD00010965
  • Molecular Weight:63.55 g/mol
  • Merck Index:13,02545
  • Boiling Pt:2595 °C (1013 hPa)
  • Melting Pt:1083 °C
  • Density:8.94 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-159-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications