Benzaldehyde, pure
Glass bottle, 500 mL
numer katalogowy CHMP421615306.0500
Dostawca: CHEMPUR
Przejdź do:
Product Details & Documents
- Numer CAS:100-52-7
- Wzór:C₆H₅CHO
- Numer MDL:MFCD00003299
- Molecular Weight:106.12 g/mol
- Indeks Mercka:13,01057
- Temperatura wrzenia:179 °C (1013 hPa)
- Temperatura topnienia:–26 °C
- Gęstość:1.049 g/cm³ (20 °C)
- Punkt zapalny:64 °C
- Temperatura przechowywania:Ambient
- EINECS:202-860-4
- UN:1990
- ADR:9,III
Dane techniczne
Dane techniczne
- Pack type:Glass bottle
- Pk:500 mL
Dane techniczne
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



