Order Entry
Poland
Orders LinkContactUsLinkComponent
Bromophenol blue indicator

Bromophenol blue indicator

Glass bottle for solids, 10 g
Catalog # CHMP121840408.0010
Supplier:   CHEMPUR
Bromophenol blue indicator
Catalog # CHMP121840408.0010
Supplier:  CHEMPUR
CAS Number:  115-39-9
Bromophenol blue indicator

Glass bottle for solids, 10 g

CAS Number:  115-39-9
Price
206 zł
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
  • CAS Number:115-39-9
  • Formula:C₁₉H₁₀Br₄O₅S
  • MDL Number:MFCD00005875
  • Molecular Weight:669.97 g/mol
  • Merck Index:13,01429
  • Melting Pt:273 °C
  • Density:1 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:204-086-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.