Order Entry
Poland
Orders LinkContactUsLinkComponent
Copper(II) carbonate basic, pure analytical reagent

Copper(II) carbonate basic, pure analytical reagent

1 kg
Catalog # CHMP116586300.1000
Irritant
Supplier:   CHEMPUR
Copper(II) carbonate basic, pure analytical reagent
Catalog # CHMP116586300.1000
Supplier:  CHEMPUR
CAS Number:  12069-69-1
Copper(II) carbonate basic, pure analytical reagent

1 kg

CAS Number:  12069-69-1
Price
316 zł
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Copper (II) carbonate dihydroxide, Copper(II) carbonate, basic, Basic copper carbonate, Copper (II) carbonate-copper (II) hydroxide (1:1)
  • CAS Number:12069-69-1
  • Formula:CuCO₃·Cu(OH)₂
  • MDL Number:MFCD00010976
  • Molecular Weight:221.12 g/mol
  • Merck Index:13,02658
  • Melting Pt:200 °C
  • Density:4 g/cm³ (20 °C)
  • EINECS:235-113-6
  • UN:3288
  • ADR:6.1,III
Specifications

Specifications

  • Pk:
    1 kg

Specifications