Order Entry
Poland
Orders LinkContactUsLinkComponent
 

FORMALDEHYDE 10% SOLUTION BUFFERED 1 * 500 mL

Catalog # CHMP114321730.0500
Supplier:   CHEMPUR
FORMALDEHYDE 10% SOLUTION BUFFERED 1 * 500 mL
Catalog # CHMP114321730.0500
Supplier:  CHEMPUR
FORMALDEHYDE 10% SOLUTION BUFFERED 1 * 500 mL
Price
26,80 zł
Log in for availability
1 * 500 mL
Product Details & Documents
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.