Order Entry
Poland
Orders LinkContactUsLinkComponent
ETHANOL 80% SOLUTION 1 * 500 mL

ETHANOL 80% SOLUTION 1 * 500 mL

Catalog # CHMP113964805.0500
Supplier:   CHEMPUR
ETHANOL 80% SOLUTION 1 * 500 mL
Catalog # CHMP113964805.0500
Supplier:  CHEMPUR
CAS Number:  
ETHANOL 80% SOLUTION 1 * 500 mL
CAS Number:  
Price
159 zł
Log in for availability
1 * 500 mL
Product Details & Documents
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.