Order Entry
Poland
Orders LinkContactUsLinkComponent
Methyl red indicator

Methyl red indicator

Glass bottle for solids, 50 g
Catalog # CHMP212725700.0050
Supplier:   CHEMPUR
Methyl red indicator
Catalog # CHMP212725700.0050
Supplier:  CHEMPUR
CAS Number:  493-52-7
Methyl red indicator

Glass bottle for solids, 50 g

CAS Number:  493-52-7
Price
263 zł
Log in for availability
1 * 50 g
Product Details & Documents
Synonyms:  2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
  • CAS Number:493-52-7
  • Formula:C₁₅H₁₅N₃O₂
  • MDL Number:MFCD00002425
  • Molecular Weight:269.3 g/mol
  • Merck Index:12,06199
  • Melting Pt:178…182 °C
  • Density:1.31 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:207-776-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications