Order Entry
Poland
Orders LinkContactUsLinkComponent
Methyl red indicator

Methyl red indicator

Glass bottle for solids, 5 g
Catalog # CHMP212725700.0005
Supplier:   CHEMPUR
Methyl red indicator
Catalog # CHMP212725700.0005
Supplier:  CHEMPUR
CAS Number:  493-52-7
Methyl red indicator

Glass bottle for solids, 5 g

CAS Number:  493-52-7
Price
50,50 zł
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
  • CAS Number:493-52-7
  • Formula:C₁₅H₁₅N₃O₂
  • MDL Number:MFCD00002425
  • Molecular Weight:269.3 g/mol
  • Merck Index:12,06199
  • Melting Pt:178…182 °C
  • Density:1.31 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:207-776-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.