Order Entry
Poland
Orders LinkContactUsLinkComponent
Benzaldehyde, pure analytical reagent

Benzaldehyde, pure analytical reagent

Glass bottle, 250 mL
Catalog # CHMP111615307.0250
Irritant
Supplier:   CHEMPUR
Benzaldehyde, pure analytical reagent
Catalog # CHMP111615307.0250
Supplier:  CHEMPUR
CAS Number:  100-52-7
Benzaldehyde, pure analytical reagent
Benzaldehyde, pure analytical reagent

Glass bottle, 250 mL

CAS Number:  100-52-7
Price
136 zł
Log in for availability
1 * 250 mL
Product Details & Documents
  • CAS Number:100-52-7
  • Formula:C₆H₅CHO
  • MDL Number:MFCD00003299
  • Molecular Weight:106.12 g/mol
  • Merck Index:13,01057
  • Boiling Pt:179 °C (1013 hPa)
  • Melting Pt:–26 °C
  • Density:1.049 g/cm³ (20 °C)
  • Flash Pt:64 °C
  • Storage Temperature:Ambient
  • EINECS:202-860-4
  • UN:1990
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.