Benzaldehyde, pure analytical reagent
Glass bottle, 100 mL
CHMP111615307.0100
: CHEMPUR
- :100-52-7
- :C₆H₅CHO
- :MFCD00003299
- :106.12 g/mol
- :13,01057
- :179 °C (1013 hPa)
- :–26 °C
- :1.049 g/cm³ (20 °C)
- :64 °C
- :Ambient
- :202-860-4
- :1990
- :9,III
- Pack type:Glass bottle
- Pk:100 mL