Order Entry
Poland
Orders LinkContactUsLinkComponent
Ammonia solution 20%, pure
Ammonia solution 20%, pure
5 L
  CHMP421349632.5000
 :   CHEMPUR
Ammonia solution 20%, pure
  CHMP421349632.5000
 :  CHEMPUR
 :  1336-21-6
Ammonia solution 20%, pure
5 L
 :  1336-21-6
189 zł
1 * 5 L
 :  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  •  :1336-21-6
  •  :NH₃
  •  :MFCD00066650
  •  :35.05 g/mol
  •  :0.92 g/cm³ (20 °C)
  •  :Ambient
  •  :215-647-6
  •  :2672
  •  :8,III
  • Pk:
    5 L