Order Entry
Poland
Orders LinkContactUsLinkComponent
Ammonia solution 10%, pure

Ammonia solution 10%, pure

Glass bottle, 500 mL
Catalog # CHMP421349604.0500
Corrosive
Irritant
Environment
Supplier:   CHEMPUR
Ammonia solution 10%, pure
Catalog # CHMP421349604.0500
Supplier:  CHEMPUR
CAS Number:  1336-21-6
Ammonia solution 10%, pure

Glass bottle, 500 mL

CAS Number:  1336-21-6
Price
31,70 zł
Log in for availability
1 * 500 mL
Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Density:0.92 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.