Order Entry
Poland
Orders LinkContactUsLinkComponent
Ammonia solution 25%, Technical Grade

Ammonia solution 25%, Technical Grade

60 L
Catalog # CHMP143618113.9060
Corrosive
Irritant
Environment
Supplier:   CHEMPUR
Ammonia solution 25%, Technical Grade
Catalog # CHMP143618113.9060
Supplier:  CHEMPUR
CAS Number:  1336-21-6
Ammonia solution 25%, Technical Grade

60 L

CAS Number:  1336-21-6
Price
1 860 zł
Log in for availability
1 * 60 L
Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Boiling Pt:32…37.7 °C (1013 hPa)
  • Melting Pt:–60…–57.5 °C
  • Density:0.88…0.91 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Specifications

Specifications

  • Pk:
    60 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.