Order Entry
Poland
Orders LinkContactUsLinkComponent
2,5-Dibromohydroquinone ≥97%

2,5-Dibromohydroquinone ≥97%

Glass bottle for solids, 25 g
Catalog # H55453.14
Irritant
2,5-Dibromohydroquinone ≥97%
Catalog # H55453.14
CAS Number:  14753-51-6
2,5-Dibromohydroquinone ≥97%
2,5-Dibromohydroquinone ≥97%

Glass bottle for solids, 25 g

CAS Number:  14753-51-6
Price
2 000 zł
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 97% CAS No.: 14753-51-6 Molecular Formula: C6H4Br2O2 Formula Weight: 267.90 Melting Point: 191-194° MDL No.: MFCD00192664

Synonyms:  2,5-Dibromobenzene-1,4-diol
  • CAS Number:14753-51-6
  • Formula:C₆H₄Br₂O₂
  • MDL Number:MFCD00192664
  • Molecular Weight:267.9 g/mol
  • Melting Pt:191…194 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.