Order Entry
Poland
Orders LinkContactUsLinkComponent
FORMALDEHYDE 10% SOLUTION BUFFERED 1 * 1 l | Avantor

FORMALDEHYDE 10% SOLUTION BUFFERED 1 * 1 l

numer katalogowy CHMP114321730.1000
Dostawca:   CHEMPUR
FORMALDEHYDE 10% SOLUTION BUFFERED 1 * 1 l
numer katalogowy CHMP114321730.1000
Dostawca:  CHEMPUR
FORMALDEHYDE 10% SOLUTION BUFFERED 1 * 1 l
Price
38,90 zł
Zaloguj się dla dostępności
1 * 1 L
Product Details & Documents
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.