Order Entry
Poland
ContactUsLinkComponent
Recombinant protein, thap2( np_113623.1, 1 a.a. - 228 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00083591-P01
Supplier:  Abnova
Recombinant protein, thap2( np_113623.1, 1 a.a. - 228 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00083591-P01
Supplier:  Abnova