Order Entry
Poland
ContactUsLinkComponent
Recombinant protein, plxdc2( np_116201.7, 29 a.a. - 108 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00084898-Q01
Supplier:  Abnova
Recombinant protein, plxdc2( np_116201.7, 29 a.a. - 108 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00084898-Q01
Supplier:  Abnova