SLC22A9 RECOMBINANT PROTEIN (Q01) 1 * 10 µG
Catalog # ABNOH00114571-Q01
Supplier: Abnova
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
Folia uszczelniająca do aplikacji ręcznej, Parafilm® M
Supplier: HEATHROW
VWR®, Rękawice nitrylowe, NITRILE
Supplier: VWR Collection
VWR® Chusteczki do twarzy
Supplier: VWR Collection
Pask wskaśnikowe pH, MQuant®, Supelco®
Supplier: Merck
Acetonitryl ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Metanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Woda, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Rękawice nitrylowe, Kimtech™ Purple Nitrile™, Kimberly-Clark
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Pipety Pasteura, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Rękawice nitrylowe
Supplier: VWR Collection