Order Entry
Poland
ContactUsLinkComponent
Recombinant protein, znf613( np_001026891.1, 1 a.a. - 617 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00079898-P01
Supplier:  Abnova
Recombinant protein, znf613( np_001026891.1, 1 a.a. - 617 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00079898-P01
Supplier:  Abnova