Recombinant protein, fbxw7( np_361014, 599 a.a. - 707 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00055294-Q01
Dostawca: Abnova
Recombinant protein, fbxw7( np_361014, 599 a.a. - 707 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00055294-Q01
Dostawca: Abnova
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Dostawca: VWR Chemicals
Sealing film for manual application, Parafilm® M
Dostawca: HEATHROW
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dostawca: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Dostawca: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Dostawca: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Dostawca: VWR Collection
VWR® Serological Pipette, Standard Line
Dostawca: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Dostawca: KIMBERLY CLARK
VWR®, Nitrile Gloves
Dostawca: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dostawca: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Dostawca: Merck
VWR® Facial Tissues
Dostawca: VWR Collection