Order Entry
Poland
ContactUsLinkComponent
Recombinant protein, kiaa0746( np_056002.1, 1 a.a. - 979 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00023231-P01
Supplier:  Abnova
Recombinant protein, kiaa0746( np_056002.1, 1 a.a. - 979 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00023231-P01
Supplier:  Abnova