Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Recombinant protein, appl1( np_036228.1, 1 a.a. - 709 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00026060-P01
Supplier:
Abnova
Expand image
Recombinant protein, appl1( np_036228.1, 1 a.a. - 709 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00026060-P01
Supplier:
Abnova
Frequently Bought Together
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
Serological pipettes, Stripette™
Supplier: Corning
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection