Order Entry
Poland
ContactUsLinkComponent
Recombinant protein, mtss1( aah23998, 346 a.a. - 445 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00009788-Q01
Supplier:  Abnova
Recombinant protein, mtss1( aah23998, 346 a.a. - 445 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00009788-Q01
Supplier:  Abnova