Order Entry
Poland
ContactUsLinkComponent
Recombinant protein, skb1( np_006100.2, 531 a.a. - 637 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00010419-Q01
Supplier:  Abnova
Recombinant protein, skb1( np_006100.2, 531 a.a. - 637 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00010419-Q01
Supplier:  Abnova