Order Entry
Poland
ContactUsLinkComponent
Recombinant protein, pcmt1( np_005380, 117 a.a. - 225 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00005110-Q01
Supplier:  Abnova
Recombinant protein, pcmt1( np_005380, 117 a.a. - 225 a.a.) partial recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00005110-Q01
Supplier:  Abnova