Order Entry
Poland
ContactUsLinkComponent
Recombinant protein, hps1( np_000186.2, 96 a.a. - 200 a.a.) partial recombinant protein with gst tag. 1 * 2 µG
Catalog # ABNOH00003257-Q01
Supplier:  Abnova
Recombinant protein, hps1( np_000186.2, 96 a.a. - 200 a.a.) partial recombinant protein with gst tag. 1 * 2 µG
Catalog # ABNOH00003257-Q01
Supplier:  Abnova